Assay Details
Target Gene Details
Entrez Gene ID: | 22192 |
Gene Name: | ubiquitin-conjugating enzyme E2M |
Gene Aliases: |
2510040H03Rik, UBC12, Ubc-rs2 |
Location: |
Chr.7:13035120-13038275 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2m | NM_001168469.2 | NP_001161941.1 | ||
| NM_001243968.1 | NP_001230897.1 | |||
| NM_145578.3 | NP_663553.1 | |||
| AK195373.1 | ||||
| BC021792.1 | AAH21792.1 | |||
| CK619787.1 |
More Information
Set Membership: |
|

Genomic Map