Assay Details
Target Gene Details
Entrez Gene ID: | 68742 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
1110032O16Rik, 2900045G02Rik, IGFBP-3R |
Location: |
Chr.7:126886219-126928680 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem219 | NM_026827.1 | NP_081103.1 | ||
| NM_028389.1 | NP_082665.1 | |||
| AK013646.1 | BAB28936.1 | |||
| AK152526.1 | BAE31285.1 | |||
| BC023442.1 | AAH23442.1 | |||
| BY128578.1 |
More Information
Set Membership: |
|

Genomic Map