Assay Details
Target Gene Details
Entrez Gene ID: | 68742 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
1110032O16Rik, 2900045G02Rik, IGFBP-3R |
Location: |
Chr.7:126886219-126928680 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem219 | NM_026827.1 | 3 | 317 | NP_081103.1 |
| NM_028389.1 | 3 | 314 | NP_082665.1 | |
| AK004043.1 | 2 | 277 | BAB23139.1 | |
| AK013646.1 | 2 | 119 | BAB28936.1 | |
| AK152526.1 | 3 | 279 | BAE31285.1 | |
| BC023442.1 | 3 | 292 | AAH23442.1 | |
| BC046763.1 | 2 | 459 | AAH46763.1 | |
| BY128578.1 |

Genomic Map