Assay Details
Target Gene Details
Entrez Gene ID: | 381921 |
Gene Name: | TAO kinase 2 |
Gene Aliases: |
1110033K02Rik, B230344N16, MAP3K17, PSK, PSK1, TAO1, TAO2, mKIAA0881 |
Location: |
Chr.7:126865677-126886296 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taok2 |
Target Gene Details
Entrez Gene ID: | 68742 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
1110032O16Rik, 2900045G02Rik, IGFBP-3R |
Location: |
Chr.7:126886219-126928680 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem219 | NM_026827.1 | NP_081103.1 | ||
| NM_028389.1 | NP_082665.1 | |||
| AK004043.1 | BAB23139.1 | |||
| AK152526.1 | BAE31285.1 | |||
| BC023442.1 | AAH23442.1 | |||
| BC046763.1 | AAH46763.1 | |||
| BP769695.1 |

Genomic Map