Assay Details
Target Gene Details
Entrez Gene ID: | 20924 |
Gene Name: | suppressor of Ty 5 |
Gene Aliases: |
AL033283, AL033321, AU019549, Spt5, Supt5h |
Location: |
Chr.7:28314891-28338804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Supt5 | NM_013676.1 | NP_038704.1 | ||
| AK145117.1 | BAE26244.1 | |||
| AK145465.1 | ||||
| AK146055.1 | BAE26864.1 | |||
| AK146453.1 | BAE27184.1 | |||
| AK146583.1 | BAE27278.1 | |||
| AK146650.1 | BAE27330.1 | |||
| AK159786.1 | ||||
| AK182624.1 | ||||
| BC007132.1 | AAH07132.1 | |||
| BC057449.1 | AAH57449.1 | |||
| BC058598.1 | AAH58598.1 | |||
| BC059849.1 | AAH59849.1 | |||
| U88539.1 | AAC40052.1 |

Genomic Map