Assay Details
Target Gene Details
Entrez Gene ID: | 170776 |
Gene Name: | CD209c antigen |
Gene Aliases: |
SIGNR2, mSIGNR2 |
Location: |
Chr.8:3940221-3955947 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd209c | NM_130903.3 | NP_570973.1 | ||
| AF373410.1 | AAL13236.1 | |||
| BC120804.1 | ||||
| BC132635.1 | ||||
| BC145088.1 |
More Information
Set Membership: |
|

Genomic Map