Assay Details
Target Gene Details
Entrez Gene ID: | 67914 |
Gene Name: | coenzyme Q9 homolog (yeast) |
Gene Aliases: |
2310005O14Rik, C78387 |
Location: |
Chr.8:94838417-94854895 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coq9 | NM_026452.2 | NP_080728.1 | ||
| AK004527.1 | BAB23347.1 | |||
| AK009179.1 | ||||
| AK154343.1 | BAE32528.1 | |||
| AK158979.1 | BAE34754.1 | |||
| BC036386.1 | AAH36386.1 |

Genomic Map