Assay Details
Target Gene Details
Entrez Gene ID: | 109006 |
Gene Name: | cytokine induced apoptosis inhibitor 1 |
Gene Aliases: |
2810413N20Rik, AA617265, AU021794, anamorsin |
Location: |
Chr.8:94819818-94838340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ciapin1 | NM_134141.4 | NP_598902.1 | ||
| AK076116.1 | BAC36196.1 | |||
| AK079615.1 | BAC37702.1 | |||
| AK088757.1 | BAC40550.1 | |||
| AK136821.1 | BAE23137.1 | |||
| AK146282.1 | BAE27040.1 | |||
| AK163705.1 | BAE37466.1 | |||
| AK168252.1 | BAE40202.1 | |||
| AK201030.1 | ||||
| AY523555.1 | AAS09959.1 | |||
| BC016261.1 | AAH16261.1 | |||
| BC021864.1 | AAH21864.1 | |||
| BC021949.1 | AAH21949.1 |

Genomic Map