Assay Details
Target Gene Details
Entrez Gene ID: | 75773 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
4930403J07Rik |
Location: |
Chr.8:119612710-119616926 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adad2 | NM_029428.1 | 2 | 415 | NP_083704.1 |
| AK015063.1 | 2 | 415 | BAB29697.1 | |
| BC137980.1 | 2 | 402 |
More Information
Set Membership: |
|

Genomic Map