Assay Details
Target Gene Details
Entrez Gene ID: | 67177 |
Gene Name: | chromatin licensing and DNA replication factor 1 |
Gene Aliases: |
2610318F11Rik, AW545653, C76791, Ris2 |
Location: |
Chr.8:122568015-122573130 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdt1 | NM_026014.3 | NP_080290.3 | ||
| AB086655.1 | BAC22085.1 | |||
| AF477481.1 | AAL82630.1 | |||
| AK012041.1 | ||||
| AK017609.1 | ||||
| AK028287.1 | BAC25859.1 | |||
| AK049163.1 | BAC33579.1 | |||
| AK076527.1 | ||||
| AK083015.1 | BAC38731.1 | |||
| BC024485.1 | AAH24485.1 | |||
| BC048076.1 | AAH48076.1 |
More Information
Set Membership: |
|

Genomic Map