Assay Details
Target Gene Details
Entrez Gene ID: | 12931 |
Gene Name: | cytokine receptor-like factor 1 |
Gene Aliases: |
CLF-1, CRLM3, NR6.1 |
Location: |
Chr.8:70493107-70504081 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crlf1 | NM_018827.2 | 4 | 741 | NP_061297.1 |
| AB040038.1 | 4 | 741 | BAA92777.1 | |
| BC107376.1 | AAI07377.1 | |||
| BC107377.1 | AAI07378.1 |
More Information
Set Membership: |
|

Genomic Map