Assay Details
Target Gene Details
Entrez Gene ID: | 104158 |
Gene Name: | carboxylesterase 1D |
Gene Aliases: |
Ces3, TGH |
Location: |
Chr.8:93166072-93197804 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1d | NM_053200.2 | 1 | 70 | NP_444430.2 |
| AB023631.1 | BAB60698.1 | |||
| AF378751.1 | 1 | 45 | AAK58067.1 | |
| AK078879.1 | 1 | 48 | BAC37439.1 | |
| BC019198.1 | AAH19198.1 | |||
| CJ133999.1 | 1 | 71 |
More Information
Set Membership: |
|

Genomic Map