Assay Details
Target Gene Details
Entrez Gene ID: | 104158 |
Gene Name: | carboxylesterase 1D |
Gene Aliases: |
Ces3, TGH |
Location: |
Chr.8:93166072-93197804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1d | NM_053200.2 | NP_444430.2 | ||
| AA522194.1 | ||||
| AB023631.1 | BAB60698.1 | |||
| AF378751.1 | AAK58067.1 | |||
| AK078879.1 | BAC37439.1 | |||
| BC019198.1 | AAH19198.1 |
More Information
Set Membership: |
|

Genomic Map