Assay Details
Target Gene Details
Entrez Gene ID: | 234776 |
Gene Name: | ATM interactor |
Gene Aliases: |
Asciz, gpg6, mKIAA0431 |
Location: |
Chr.8:116943386-116960445 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atmin | NM_177700.4 | 3 | 585 | NP_808368.3 |
| AK122281.1 | 3 | 467 | BAC65563.1 | |
| AX775769.1 | 3 | 585 | CAE11584.1 | |
| BC027752.1 | 3 | 559 | ||
| BC060631.1 | 3 | 570 | AAH60631.1 |

Genomic Map