Assay Details
Target Gene Details
Entrez Gene ID: | 20299 |
Gene Name: | chemokine (C-C motif) ligand 22 |
Gene Aliases: |
ABCD-1, DCBCK, MDC, Scya22 |
Location: |
Chr.8:94745684-94751388 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccl22 | NM_009137.2 | NP_033163.1 | ||
| AF052505.1 | 1 | 144 | AAC40200.1 | |
| AF076596.1 | AAD50547.1 | |||
| AF163476.1 | AAD55763.1 | |||
| AJ238238.1 | CAB40894.1 | |||
| AK155083.1 | BAE33035.1 | |||
| AK155764.1 | BAE33426.1 | |||
| AK156627.1 | BAE33782.1 | |||
| AK170080.1 | BAE41551.1 | |||
| BC012658.1 | AAH12658.1 | |||
| BY198556.1 |
More Information
Set Membership: |
|

Genomic Map