Assay Details
Target Gene Details
Entrez Gene ID: | 30947 |
Gene Name: | adenosine deaminase, tRNA-specific 1 |
Gene Aliases: |
MMADAT1, mADAT1 |
Location: |
Chr.8:111966897-111992307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adat1 | NM_013925.4 | NP_038953.1 | ||
| AF192375.1 | AAF36820.1 | |||
| AK035614.1 | 7 | 1265 | BAC29125.1 | |
| AK039699.1 | BAC30422.1 | |||
| AK082399.1 | BAC38487.1 | |||
| AK161044.1 | 9 | 1493 | BAE36167.1 | |
| BC019976.1 | 9 | 1469 | AAH19976.1 |
More Information
Set Membership: |
|

Genomic Map