Assay Details
Target Gene Details
Entrez Gene ID: | 69225 |
Gene Name: | carbohydrate kinase domain containing |
Gene Aliases: |
0710008K08Rik, 2810407E01Rik |
Location: |
Chr.8:11497506-11513287 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Carkd | NM_001190357.1 | NP_001177286.1 | ||
| NM_001293661.1 | NP_001280590.1 | |||
| NM_026995.4 | 1 | 53 | NP_081271.2 | |
| AK003048.1 | BAB22531.1 | |||
| AK009254.1 | BAB26172.1 | |||
| AK012456.1 | BAB28251.1 | |||
| AK013028.1 | BAB28607.1 | |||
| AK013069.1 | BAB28632.1 | |||
| AK013420.1 | BAB28847.1 | |||
| AK039642.1 | 1 | 35 | ||
| AK154142.1 | 1 | 35 | BAE32403.1 | |
| AK171034.1 | 1 | 35 | BAE42200.1 | |
| BC019538.1 | AAH19538.1 | |||
| BC021955.1 | AAH21955.1 | |||
| BY254286.1 | 1 | 54 | ||
| BY270677.1 | ||||
| DV045296.1 |
More Information
Set Membership: |
|

Genomic Map