Assay Details
Target Gene Details
Entrez Gene ID: | 234594 |
Gene Name: | CCR4-NOT transcription complex, subunit 1 |
Gene Aliases: |
6030411K04Rik, AA815922, D830048B13 |
Location: |
Chr.8:95719451-95807479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 48 - Exon 48 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot1 | NM_001205226.1 | NP_001192155.1 | ||
| NM_153164.3 | NP_694804.3 | |||
| NM_178078.2 | NP_835179.1 | |||
| AK031357.1 | BAC27364.1 | |||
| AK048177.1 | 8 | 2774 | BAC33267.1 | |
| AK079464.1 | ||||
| BC018281.1 | ||||
| BC094620.1 | AAH94620.1 | |||
| BC157948.1 | ||||
| BC158073.1 | ||||
| BC172105.1 |
Target Gene Details
Entrez Gene ID: | 66083 |
Gene Name: | SET domain containing 6 |
Gene Aliases: |
0610039J04Rik, 3110004G14Rik, AI413388, C76402 |
Location: |
Chr.8:95715113-95721499 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd6 |
More Information
Set Membership: |
|

Genomic Map