Assay Details
Target Gene Details
Entrez Gene ID: | 319960 |
Gene Name: | RIKEN cDNA 4930513N10 gene |
Gene Aliases: |
- |
Location: |
Chr.8:95806830-95821728 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930513N10Rik | NR_015574.2 | 1 | 654 | |
| AK029729.1 | 1 | 512 | ||
| AK042481.1 | 1 | 740 | ||
| AK147860.1 | 1 | 655 | ||
| AK161453.1 | 1 | 509 | BAE36405.1 |
Target Gene Details
Entrez Gene ID: | 234594 |
Gene Name: | CCR4-NOT transcription complex, subunit 1 |
Gene Aliases: |
6030411K04Rik, AA815922, D830048B13 |
Location: |
Chr.8:95719451-95807479 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot1 | NM_001205226.1 | NP_001192155.1 | ||
| NM_153164.3 | NP_694804.3 | |||
| NM_178078.2 | NP_835179.1 | |||
| AK086041.1 | ||||
| AK169894.1 |
More Information
Set Membership: |
|

Genomic Map