Assay Details
Target Gene Details
Entrez Gene ID: | 414077 |
Gene Name: | WD repeat domain 83 opposite strand |
Gene Aliases: |
- |
Location: |
Chr.8:85080963-85082339 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr83os | NM_001001493.2 | NP_001001493.1 | ||
| AK010893.1 | BAB27250.1 | |||
| AK147129.1 | BAE27699.1 | |||
| BC056474.1 | AAH56474.1 | |||
| BC089623.1 | AAH89623.1 |

Genomic Map