Assay Details
Target Gene Details
Entrez Gene ID: | 234594 |
Gene Name: | CCR4-NOT transcription complex, subunit 1 |
Gene Aliases: |
6030411K04Rik, AA815922, D830048B13 |
Location: |
Chr.8:95719451-95807479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot1 | NM_001205226.1 | NP_001192155.1 | ||
| NM_153164.3 | NP_694804.3 | |||
| NM_178078.2 | NP_835179.1 | |||
| AK034776.1 | BAC28830.1 | |||
| AK143651.1 | BAE25479.1 | |||
| BC035210.1 | AAH35210.1 | |||
| BC157948.1 | ||||
| BC158073.1 | ||||
| BC172105.1 |
Target Gene Details
Entrez Gene ID: | 102465650 |
Gene Name: | microRNA 7073 |
Gene Aliases: |
mmu-mir-7073 |
Location: |
Chr.8:95753151-95753214 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7073 | NR_106041.1 |
More Information
Set Membership: |
|

Genomic Map