Assay Details
Target Gene Details
Entrez Gene ID: | 20312 |
Gene Name: | chemokine (C-X3-C motif) ligand 1 |
Gene Aliases: |
AB030188, ABCD-3, AI848747, CX3C, Cxc3, D8Bwg0439e, Scyd1 |
Location: |
Chr.8:94772180-94782427 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cx3cl1 | NM_009142.3 | NP_033168.2 | ||
| AB030188.1 | 1 | 133 | BAA92752.1 | |
| AF010586.1 | AAB66331.1 | |||
| AF071549.1 | AAD41620.1 | |||
| AK040220.1 | BAC30544.1 | |||
| AK078285.1 | ||||
| AK138214.1 | BAE23585.1 | |||
| AK138472.1 | BAE23673.1 | |||
| AK138574.1 | BAE23702.1 | |||
| AK154646.1 | BAE32737.1 | |||
| BC006650.1 | AAH06650.1 | |||
| BC054838.1 | AAH54838.1 | |||
| CB526167.1 | ||||
| U92565.1 | AAB71763.1 |
More Information
Set Membership: |
|

Genomic Map