Assay Details
Target Gene Details
Entrez Gene ID: | 56445 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily A, member 2 |
Gene Aliases: |
1500017M13Rik, 2010206B19Rik, DNAJ, DNJ3, Dnaj3, HIRIP4, PRO3015, mDj3 |
Location: |
Chr.8:85537640-85555271 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaja2 | NM_019794.4 | NP_062768.1 | ||
| AB028853.1 | BAA88301.1 | |||
| AK031858.1 | ||||
| AK077672.1 | BAC36946.1 | |||
| AK083208.1 | BAC38809.1 | |||
| AK163255.1 | ||||
| AK169169.1 | BAE40948.1 | |||
| BC003420.1 | AAH03420.1 |
More Information
Set Membership: |
|

Genomic Map