Assay Details
Target Gene Details
Entrez Gene ID: | 234686 |
Gene Name: | formin homology 2 domain containing 1 |
Gene Aliases: |
BC017144, FHOS, FHOS1 |
Location: |
Chr.8:105329160-105347970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhod1 | NM_177699.4 | NP_808367.2 | ||
| AB041045.1 | BAE06182.1 | |||
| AK030737.1 | BAC27106.1 | |||
| BC060654.1 | AAH60654.1 |
Target Gene Details
Entrez Gene ID: | 66320 |
Gene Name: | transmembrane protein 208 |
Gene Aliases: |
1700006C06Rik, 2610030K20Rik, Hspc171 |
Location: |
Chr.8:105326364-105335485 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem208 | AK131833.1 | BAE20823.1 |

Genomic Map