Assay Details
Target Gene Details
Entrez Gene ID: | 76615 |
Gene Name: | glutamic-oxaloacetic transaminase 1-like 1 |
Gene Aliases: |
1700083M11Rik |
Location: |
Chr.8:27197459-27223848 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Got1l1 | NM_029674.1 | NP_083950.1 | ||
| BC052754.1 | AAH52754.1 |
More Information
Set Membership: |
|

Genomic Map