Assay Details
Target Gene Details
Entrez Gene ID: | 16795 |
Gene Name: | like-glycosyltransferase |
Gene Aliases: |
BPFD#36, Gyltl1a, Mbp-1, Mbp1, enr, fg, froggy, mKIAA0609, myd |
Location: |
Chr.8:72814599-73353577 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Large | NM_010687.1 | NP_034817.1 | ||
| AJ006278.1 | CAA06945.1 | |||
| AK122328.1 | BAC65610.1 | |||
| AK133940.1 | ||||
| BC061506.1 | AAH61506.1 | |||
| BC100399.1 | AAI00400.1 | |||
| BC125487.1 | ||||
| BC125489.1 | ||||
| HM141820.1 |
More Information
Set Membership: |
|

Genomic Map