Assay Details
Target Gene Details
Entrez Gene ID: | 100503676 |
Gene Name: | predicted gene, 38426 |
Gene Aliases: |
- |
Location: |
Chr.8:84913335-84922915 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm38426 | NR_103491.1 | |||
| AI527494.1 | ||||
| AK018651.1 |
Target Gene Details
Entrez Gene ID: | 56527 |
Gene Name: | microtubule associated serine/threonine kinase 1 |
Gene Aliases: |
9430008B02Rik, SAST, SAST170 |
Location: |
Chr.8:84911853-84946129 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mast1 | NM_019945.2 | NP_064329.2 | ||
| AF077818.1 | AAD50548.1 | |||
| AK122411.1 | BAC65693.3 | |||
| AK141034.1 | BAE24549.1 | |||
| BC054524.1 | AAH54524.1 |
More Information
Set Membership: |
|

Genomic Map