Assay Details
Target Gene Details
Entrez Gene ID: | 16150 |
Gene Name: | inhibitor of kappaB kinase beta |
Gene Aliases: |
AI132552, IKK-2, IKK-beta, IKK2, IKK[b], IKKbeta |
Location: |
Chr.8:22659205-22706585 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkb | NM_001159774.1 | 19 | 2049 | NP_001153246.1 |
| NM_010546.2 | 19 | 2049 | NP_034676.1 | |
| AF026524.1 | 18 | 1917 | AAC23557.1 | |
| AF088910.1 | 19 | 2109 | AAD52095.1 | |
| AK144603.1 | 19 | 2042 | BAE25962.1 | |
| AK150884.1 | 18 | 2171 | BAE29931.1 | |
| AK151888.1 | 18 | 2171 | ||
| AK151955.1 | 19 | 2001 | BAE30827.1 | |
| AK153151.1 | 19 | 2001 | BAE31761.1 | |
| AK155587.1 | 17 | 4400 | ||
| AK156294.1 | 19 | 2002 | BAE33659.1 | |
| AK219506.1 | ||||
| BC004772.1 | 19 | 1987 | ||
| BC037723.1 | 19 | 2021 | AAH37723.1 |
More Information
Set Membership: |
|

Genomic Map