Assay Details
Target Gene Details
Entrez Gene ID: | 16150 |
Gene Name: | inhibitor of kappaB kinase beta |
Gene Aliases: |
AI132552, IKK-2, IKK-beta, IKK2, IKK[b], IKKbeta |
Location: |
Chr.8:22659205-22706585 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkb | NM_001159774.1 | NP_001153246.1 | ||
| NM_010546.2 | NP_034676.1 | |||
| AF026524.1 | AAC23557.1 | |||
| AF088910.1 | AAD52095.1 | |||
| AK011359.1 | 7 | 707 | BAE43224.1 | |
| AK086442.1 | ||||
| AK139972.1 | BAE24202.1 | |||
| AK144603.1 | BAE25962.1 | |||
| AK150884.1 | BAE29931.1 | |||
| AK151888.1 | ||||
| AK151955.1 | BAE30827.1 | |||
| AK153151.1 | BAE31761.1 | |||
| AK155587.1 | ||||
| AK156294.1 | BAE33659.1 | |||
| BC004772.1 | ||||
| BC037723.1 | AAH37723.1 |
More Information
Set Membership: |
|

Genomic Map