Assay Details
Target Gene Details
Entrez Gene ID: | 100044509 |
Gene Name: | transforming growth factor, beta receptor III-like |
Gene Aliases: |
Gm14378, OTTMUSG00000016477 |
Location: |
Chr.8:4248214-4251423 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfbr3l | NM_001195258.1 | NP_001182187.1 |
More Information
Set Membership: |
|

Genomic Map