Assay Details
Target Gene Details
Entrez Gene ID: | 50931 |
Gene Name: | interleukin 27 receptor, alpha |
Gene Aliases: |
CRL1, IL-27R, Tccr, WSX-1, Wsx1, zcytor1 |
Location: |
Chr.8:84030286-84042629 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il27ra | NM_016671.3 | NP_057880.2 | ||
| AF053005.1 | AAC40121.1 | |||
| AF265243.1 | AAG27298.1 | |||
| AK040457.1 | ||||
| AK133753.1 | BAE21823.1 | |||
| BC032878.1 | AAH32878.1 |
More Information
Set Membership: |
|

Genomic Map