Assay Details
Target Gene Details
Entrez Gene ID: | 16532 |
Gene Name: | potassium channel, subfamily U, member 1 |
Gene Aliases: |
Kcnma3, Slo3, mSlo3 |
Location: |
Chr.8:25842654-25937943 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnu1 | NM_008432.3 | NP_032458.3 | ||
| AF039213.2 | AAB99742.2 | |||
| AK083174.1 |
More Information
Set Membership: |
|

Genomic Map