Assay Details
Target Gene Details
Entrez Gene ID: | 26386 |
Gene Name: | heat shock transcription factor 4 |
Gene Aliases: |
HSF 4, HSTF 4, HSTF4, ldis1, mHSF4 |
Location: |
Chr.8:105269748-105275845 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsf4 | NM_001256042.1 | NP_001242971.1 | ||
| NM_001256044.1 | NP_001242973.1 | |||
| NM_011939.3 | NP_036069.1 | |||
| NR_045689.1 | ||||
| AB029349.1 | BAA84583.1 | |||
| AB029350.1 | BAA84584.1 | |||
| AF160966.1 | AAF80399.1 | |||
| AK148056.1 | BAE28316.1 | |||
| BC138130.1 |
More Information
Set Membership: |
|

Genomic Map