Assay Details
Target Gene Details
Entrez Gene ID: | 16532 |
Gene Name: | potassium channel, subfamily U, member 1 |
Gene Aliases: |
Kcnma3, Slo3, mSlo3 |
Location: |
Chr.8:25842654-25937943 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnu1 | NM_008432.3 | 25 | 2917 | NP_032458.3 |
| AF039213.2 | 25 | 2884 | AAB99742.2 | |
| AK083174.1 | 6 | 833 | ||
| BQ175543.1 | 3 | 632 |
More Information
Set Membership: |
|

Genomic Map