Assay Details
Target Gene Details
Entrez Gene ID: | 234421 |
Gene Name: | calcium and integrin binding family member 3 |
Gene Aliases: |
C730014M21Rik, Gm1107, KIP3 |
Location: |
Chr.8:72192699-72213092 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cib3 |
Target Gene Details
Entrez Gene ID: | 209488 |
Gene Name: | hematopoietic SH2 domain containing |
Gene Aliases: |
ALX, Hsh2 |
Location: |
Chr.8:72189668-72200958 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsh2d | NM_197944.1 | NP_922935.1 | ||
| AY319653.1 | AAQ81286.1 | |||
| BC094291.1 | AAH94291.1 |
More Information
Set Membership: |
|

Genomic Map