Assay Details
Target Gene Details
Entrez Gene ID: | 73711 |
Gene Name: | multivesicular body subunit 12A |
Gene Aliases: |
1110012M11Rik, Fam125a |
Location: |
Chr.8:71542880-71548026 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mvb12a | NM_028617.2 | NP_082893.1 | ||
| AK003644.1 | ||||
| BC020119.1 | AAH20119.1 | |||
| BC027419.1 | AAH27419.1 | |||
| BC087734.1 | AAH87734.1 |

Genomic Map