Assay Details
Target Gene Details
Entrez Gene ID: | 75311 |
Gene Name: | RIKEN cDNA 4930550C14 gene |
Gene Aliases: |
AU017961 |
Location: |
Chr.9:53401888-53438266 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930550C14Rik | NM_029247.3 | 7 | 726 | NP_083523.1 |
| AK016083.1 | 7 | 727 | BAB30107.1 | |
| AK153618.1 | 5 | 623 | BAE32121.1 | |
| BC115638.1 | 5 | 627 | ||
| BC115639.1 | 7 | 714 |
More Information
Set Membership: |
|

Genomic Map