Assay Details
Target Gene Details
Entrez Gene ID: | 69478 |
Gene Name: | RIKEN cDNA 2300009A05 gene |
Gene Aliases: |
- |
Location: |
Chr.9:63394447-63399244 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2300009A05Rik | NM_027090.1 | 2 | 449 | NP_081366.1 |
| AK009046.1 | 2 | 448 | BAB26045.1 | |
| BY090300.1 | 2 | 192 |

Genomic Map