Assay Details
Target Gene Details
Entrez Gene ID: | 244416 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3B |
Gene Aliases: |
6430576E21, AW821953, GL, PTG |
Location: |
Chr.8:35375668-35388139 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3b | NM_177741.3 | NP_808409.1 | ||
| AK052438.1 | BAC34990.1 | |||
| BC079666.1 | AAH79666.1 |
More Information
Set Membership: |
|

Genomic Map