Assay Details
Target Gene Details
Entrez Gene ID: | 69642 |
Gene Name: | muscular LMNA-interacting protein |
Gene Aliases: |
2310046A06Rik, AI931782, CIP |
Location: |
Chr.9:77102084-77347870 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlip | NM_027150.1 | NP_081426.1 | ||
| AK009836.1 | BAB26534.1 | |||
| BC089626.1 | AAH89626.1 | |||
| BC131903.1 | ||||
| BC144730.1 |

Genomic Map