Assay Details
Target Gene Details
Entrez Gene ID: | 20911 |
Gene Name: | syntaxin binding protein 2 |
Gene Aliases: |
C79054, Munc-18-2, Munc-18b, Munc18b, Sxtbp2, Sxtp2, Unc18-2, Unc18b, muSec1 |
Location: |
Chr.8:3630960-3643707 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp2 | NM_011503.4 | NP_035633.1 | ||
| AF263345.1 | AAF71615.1 | |||
| AK088364.1 | BAC40304.1 | |||
| AK133419.1 | BAE21647.1 | |||
| BC003477.1 | AAH03477.1 | |||
| BC008102.1 | AAH08102.1 | |||
| D42068.1 | BAA07666.1 | |||
| U19520.1 | AAA69912.1 |
More Information
Set Membership: |
|

Genomic Map