Assay Details
Target Gene Details
Entrez Gene ID: | 66432 |
Gene Name: | solute carrier family 7, member 6 opposite strand |
Gene Aliases: |
2010007L18Rik, 2400002F02Rik |
Location: |
Chr.8:106200438-106210992 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a6os | NM_001007567.2 | NP_001007568.1 | ||
| AK010254.1 | ||||
| AK134626.1 | ||||
| AK146733.1 | BAE27395.1 | |||
| AK171806.1 | ||||
| BC054118.1 | AAH54118.1 |

Genomic Map