Assay Details
Target Gene Details
Entrez Gene ID: | 67971 |
Gene Name: | tubulin polymerization-promoting protein family member 3 |
Gene Aliases: |
2700055K07Rik, CGI-38, Ceacam9, mmCGM8 |
Location: |
Chr.8:105467492-105471422 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tppp3 | NM_026481.3 | NP_080757.1 | ||
| AK009771.1 | BAB26493.1 | |||
| AK012437.1 | BAB28237.1 | |||
| AK155018.1 | BAE32991.1 | |||
| AK160507.1 | BAE35831.1 | |||
| BC010788.1 | AAH10788.1 | |||
| BF469439.1 |
More Information
Set Membership: |
|

Genomic Map