Assay Details
Target Gene Details
Entrez Gene ID: | 234724 |
Gene Name: | tyrosine aminotransferase |
Gene Aliases: |
- |
Location: |
Chr.8:109990436-109999804 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tat | NM_146214.3 | NP_666326.1 | ||
| AK090244.1 | BAC41146.1 | |||
| AK149383.1 | BAE28844.1 | |||
| AK149526.1 | BAE28939.1 | |||
| BC023949.1 | AAH23949.1 | |||
| BC024120.1 | AAH24120.1 | |||
| BC025934.1 | AAH25934.1 | |||
| BC028821.1 | AAH28821.1 | |||
| BC030728.1 | AAH30728.1 | |||
| BC030729.1 | AAH30729.1 | |||
| BC037526.1 | AAH37526.1 |
More Information
Set Membership: |
|

Genomic Map