Assay Details
Target Gene Details
Entrez Gene ID: | 18709 |
Gene Name: | phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta) |
Gene Aliases: |
p85beta |
Location: |
Chr.8:70768178-70776728 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r2 | NM_008841.2 | 15 | 2361 | NP_032867.2 |
| AK146195.1 | 15 | 2351 | BAE26969.1 | |
| AK148122.1 | 15 | 2358 | ||
| BC006796.1 | 15 | 2352 | AAH06796.1 | |
| BC085501.1 | 14 | 2284 | AAH85501.1 | |
| Y13569.1 | 15 | 2330 | CAA73903.1 |
More Information
Set Membership: |
|

Genomic Map