Assay Details
Target Gene Details
Entrez Gene ID: | 68051 |
Gene Name: | nuclear transport factor 2 |
Gene Aliases: |
2700067I02Rik, AI115459, AI413596, AW546000, Ntf2, Ntf3, PP15 |
Location: |
Chr.8:105860634-105880402 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nutf2 | NM_026532.3 | NP_080808.1 | ||
| AK002461.1 | BAB22117.1 | |||
| AK012500.1 | BAB28283.1 | |||
| AK020512.1 | BAB32122.1 | |||
| AK028410.1 | BAC25936.1 | |||
| AK051049.1 | BAC34511.1 | |||
| AK152212.1 | BAE31040.1 | |||
| AK168026.1 | BAE40011.1 | |||
| BC003955.1 | AAH03955.1 | |||
| BC083165.1 | AAH83165.1 |

Genomic Map