Assay Details
Target Gene Details
Entrez Gene ID: | 19358 |
Gene Name: | RAD23a homolog (S. cerevisiae) |
Gene Aliases: |
2310040P19Rik, AL024030, HR23A, mHR23A |
Location: |
Chr.8:84834652-84840750 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad23a | NM_001297606.1 | NP_001284535.1 | ||
| NM_001297607.1 | NP_001284536.1 | |||
| NM_009010.5 | NP_033036.2 | |||
| AK038300.1 | BAC29962.1 | |||
| AK045224.1 | BAC32268.1 | |||
| AK161875.1 | ||||
| AK165467.1 | BAE38204.1 | |||
| BC003846.1 | ||||
| BC138684.1 | ||||
| BC145372.1 | ||||
| X92410.1 | CAA63145.1 |
More Information
Set Membership: |
|

Genomic Map