Assay Details
Target Gene Details
Entrez Gene ID: | 234542 |
Gene Name: | retbindin |
Gene Aliases: |
A330096I21Rik, BC022744, Rtbnd |
Location: |
Chr.8:84946263-84956606 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtbdn | NM_144929.2 | NP_659178.1 | ||
| AY527275.1 | AAS20324.1 | |||
| AY527276.1 | AAS20325.1 |

Genomic Map