Assay Details
Target Gene Details
Entrez Gene ID: | 74306 |
Gene Name: | protease, serine 46 |
Gene Aliases: |
1700112C13Rik, 79H19B, Tessp6 |
Location: |
Chr.9:110844506-110856522 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss46 | NM_183103.2 | NP_898926.2 | ||
| AK007173.1 | BAC25167.1 |
More Information
Set Membership: |
|

Genomic Map