Assay Details
Target Gene Details
Entrez Gene ID: | 66083 |
Gene Name: | SET domain containing 6 |
Gene Aliases: |
0610039J04Rik, 3110004G14Rik, AI413388, C76402 |
Location: |
Chr.8:95715113-95721499 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd6 | NM_001035123.3 | NP_001030295.1 | ||
| AK010304.1 | BAB26837.1 | |||
| AK075597.1 | BAC35846.1 | |||
| AK167753.1 | BAE39788.1 | |||
| AK181998.1 | ||||
| AK215534.1 | ||||
| BC139198.1 | ||||
| BC139199.1 |
More Information
Set Membership: |
|

Genomic Map